siRNA Details
Gene Symbol: Cmpk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025655.1 | NP_001020826.1 | 3, 4 | 572 | ||
GenBank mRNA | BC098727.1 | - | 3, 4 | 572 | 7379366 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Cmpk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025655.1 | NP_001020826.1 | 3, 4 | 572 | ||
GenBank mRNA | BC098727.1 | - | 3, 4 | 572 | 7379366 |