siRNA Details
Gene Symbol: LOC103690007 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008763980.1 | XP_008762202.1 | 2079 |
Gene Symbol: Nsun4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106678.1 | NP_001100148.1 | 6 | 1185 |
