siRNA Details
| Gene Symbol: Rad54l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134960.1 | NP_001128432.1 | 4 | 579 | ||
| GenBank mRNA | BC169034.1 | - | 4 | 579 | 9089933 | |
| FQ230836.1 | - | 4 | 587 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rad54l| Gene Symbol: Rad54l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134960.1 | NP_001128432.1 | 4 | 579 | ||
| GenBank mRNA | BC169034.1 | - | 4 | 579 | 9089933 | |
| FQ230836.1 | - | 4 | 587 | |||