siRNA Details
Gene Symbol: Rad54l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134960.1 | NP_001128432.1 | 13 | 1760 | ||
GenBank mRNA | BC158840.1 | - | 5 | 512 | ||
BC169034.1 | - | 13 | 1760 | 9089933 | ||
FQ230836.1 | - | 11 | 2125 |
