siRNA Details
| Gene Symbol: Tspan1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004236.1 | NP_001004236.1 | 3 | 325 | ||
| XM_006238639.1 | XP_006238701.1 | 5 | 598 | |||
| GenBank mRNA | BC078938.1 | - | 3 | 325 | 7108356 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tspan1| Gene Symbol: Tspan1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004236.1 | NP_001004236.1 | 3 | 325 | ||
| XM_006238639.1 | XP_006238701.1 | 5 | 598 | |||
| GenBank mRNA | BC078938.1 | - | 3 | 325 | 7108356 | |