siRNA Details
| Gene Symbol: Nasp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005543.1 | NP_001005543.1 | 15 | 2329 | ||
| XM_006238646.2 | XP_006238708.1 | 15 | 2275 | |||
| XM_006238647.2 | XP_006238709.1 | 13 | 1311 | |||
| XM_008763993.1 | XP_008762215.1 | 15 | 2352 | |||
| GenBank mRNA | BC081913.1 | - | 15 | 2329 | 7112076 | |
| FQ212041.1 | - | 14 | 2269 | |||

Nasp
Genomic Map