siRNA Details
| Gene Symbol: Ppcs | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039010.2 | NP_001034099.2 | 1 | 434 | ||
| GenBank mRNA | BC099208.1 | - | 1 | 824 | 7384489 | |
| CK474267.1 | - | 1 | 473 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ppcs| Gene Symbol: Ppcs | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039010.2 | NP_001034099.2 | 1 | 434 | ||
| GenBank mRNA | BC099208.1 | - | 1 | 824 | 7384489 | |
| CK474267.1 | - | 1 | 473 | |||