siRNA Details
| Gene Symbol: Ppcs | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039010.2 | NP_001034099.2 | 3 | 667 | ||
| XM_008763998.1 | XP_008762220.1 | 3 | 452 | |||
| XM_008763999.1 | XP_008762221.1 | 3 | 237 | |||
| XM_008764000.1 | XP_008762222.1 | 2 | 375 | |||
| GenBank mRNA | BC099208.1 | - | 2 | 1435 | 7384489 | |
| BQ195046.1 | - | 1 | 702 | |||
| CK474267.1 | - | 3 | 706 | |||

Ppcs
Genomic Map