siRNA Details
| Gene Symbol: Ppcs | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039010.2 | NP_001034099.2 | 1, 2 | 511 | ||
| GenBank mRNA | CK474267.1 | - | 1, 2 | 550 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ppcs| Gene Symbol: Ppcs | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039010.2 | NP_001034099.2 | 1, 2 | 511 | ||
| GenBank mRNA | CK474267.1 | - | 1, 2 | 550 | ||