siRNA Details
| Gene Symbol: Smap2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100669.1 | NP_001094139.1 | 8 | 1170 | ||
| GenBank mRNA | BC099210.1 | - | 1 | 33 | ||
| BC161927.1 | - | 8 | 1164 | 8374170 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Smap2| Gene Symbol: Smap2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100669.1 | NP_001094139.1 | 8 | 1170 | ||
| GenBank mRNA | BC099210.1 | - | 1 | 33 | ||
| BC161927.1 | - | 8 | 1164 | 8374170 | ||