siRNA Details
| Gene Symbol: Smap2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100669.1 | NP_001094139.1 | 3 | 652 | ||
| GenBank mRNA | BC161927.1 | - | 3 | 646 | 8374170 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Smap2| Gene Symbol: Smap2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100669.1 | NP_001094139.1 | 3 | 652 | ||
| GenBank mRNA | BC161927.1 | - | 3 | 646 | 8374170 | |