siRNA Details
| Gene Symbol: Mfsd2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106683.1 | NP_001100153.1 | 3 | 398 | ||
| XM_006238792.2 | XP_006238854.1 | 3 | 378 | |||
| XM_008764001.1 | XP_008762223.1 | 3 | 375 | |||
| GenBank mRNA | BC161915.1 | - | 3 | 385 | 8369688 | |

Mfsd2a
Genomic Map