siRNA Details
| Gene Symbol: Epha10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135709.1 | NP_001129181.1 | 16, 17 | 2646 | ||
| XM_006238858.2 | XP_006238920.1 | 14, 15 | 2638 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Epha10| Gene Symbol: Epha10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135709.1 | NP_001129181.1 | 16, 17 | 2646 | ||
| XM_006238858.2 | XP_006238920.1 | 14, 15 | 2638 | |||