siRNA Details
Gene Symbol: Cnksr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039011.1 | NP_001034100.1 | 14 | 1290 | ||
GenBank mRNA | BC099788.1 | - | 14 | 1290 | 7934518 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Cnksr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039011.1 | NP_001034100.1 | 14 | 1290 | ||
GenBank mRNA | BC099788.1 | - | 14 | 1290 | 7934518 |