siRNA Details
| Gene Symbol: Mtfr1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013935.1 | NP_001013957.1 | 5 | 408 | ||
| XM_006239124.2 | XP_006239186.1 | 5 | 704 | |||
| XM_006239125.2 | XP_006239187.1 | 5 | 482 | |||
| GenBank mRNA | BC083692.1 | - | 5 | 408 | 7189448 | |

Mtfr1l
Genomic Map