siRNA Details
| Gene Symbol: Klhdc7a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006239296.2 | XP_006239358.1 | 1 | 1668 | ||
| XM_006239297.2 | XP_006239359.1 | 2 | 1552 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Klhdc7a| Gene Symbol: Klhdc7a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006239296.2 | XP_006239358.1 | 1 | 1668 | ||
| XM_006239297.2 | XP_006239359.1 | 2 | 1552 | |||