siRNA Details
Gene Symbol: Rer1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039012.1 | NP_001034101.1 | 6, 7 | 566 | ||
GenBank mRNA | BC100270.1 | - | 6, 7 | 566 | 7444634 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Rer1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039012.1 | NP_001034101.1 | 6, 7 | 566 | ||
GenBank mRNA | BC100270.1 | - | 6, 7 | 566 | 7444634 |