siRNA Details
| Gene Symbol: Atad3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034922.1 | NP_001030094.1 | 14 | 1449 | ||
| GenBank mRNA | BC105762.1 | - | 14 | 1449 | 7929859 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Atad3a| Gene Symbol: Atad3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034922.1 | NP_001030094.1 | 14 | 1449 | ||
| GenBank mRNA | BC105762.1 | - | 14 | 1449 | 7929859 | |