siRNA Details
| Gene Symbol: Aurkaip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004237.1 | NP_001004237.1 | 3 | 205 | ||
| XM_006239546.2 | XP_006239608.1 | 3 | 393 | |||
| XM_006239547.2 | XP_006239609.1 | 3 | 603 | |||
| XM_006239548.2 | XP_006239610.1 | 3 | 350 | |||
| XM_008764351.1 | XP_008762573.1 | 3 | 226 | |||
| GenBank mRNA | BC079171.1 | - | 3 | 205 | 7129954 | |
| FQ228270.1 | - | 3 | 213 | |||
| FQ232591.1 | - | 3 | 213 | |||

Aurkaip1
Genomic Map