siRNA Details
| Gene Symbol: Plekhn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001134523.1 | NP_001127995.1 | 13 | 1982 | ||
| XM_006239564.2 | XP_006239626.1 | 13 | 2213 | |||
| XM_006239565.2 | XP_006239627.1 | 13 | 2213 | |||
| XM_006239566.2 | XP_006239628.1 | 13 | 2147 | |||

Plekhn1
Genomic Map