siRNA Details
Gene Symbol: Ppan | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011980.1 | NP_001011980.1 | 5 | 444 | ||
GenBank mRNA | BC089842.1 | - | 5 | 444 | 7366262 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ppan | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011980.1 | NP_001011980.1 | 5 | 444 | ||
GenBank mRNA | BC089842.1 | - | 5 | 444 | 7366262 |