siRNA Details
| Gene Symbol: Memo1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001029917.1 | NP_001025088.1 | 3 | 301 | ||
| GenBank mRNA | BC098061.1 | - | 3 | 301 | 7458219 | |
| FQ215386.1 | - | 1 | 53 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Memo1| Gene Symbol: Memo1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001029917.1 | NP_001025088.1 | 3 | 301 | ||
| GenBank mRNA | BC098061.1 | - | 3 | 301 | 7458219 | |
| FQ215386.1 | - | 1 | 53 | |||