siRNA Details
| Gene Symbol: Atraid | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127526.1 | NP_001120998.1 | 3 | 285 | ||
| GenBank mRNA | BC166904.1 | - | 3 | 285 | 6887237 | |
| FQ214285.1 | - | 3 | 311 | |||
| FQ222311.1 | - | 3 | 286 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Atraid| Gene Symbol: Atraid | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001127526.1 | NP_001120998.1 | 3 | 285 | ||
| GenBank mRNA | BC166904.1 | - | 3 | 285 | 6887237 | |
| FQ214285.1 | - | 3 | 311 | |||
| FQ222311.1 | - | 3 | 286 | |||