siRNA Details
| Gene Symbol: Osr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106716.2 | NP_001100186.1 | 1, 2 | 707 | ||
| XM_006239886.2 | XP_006239948.1 | 2, 3 | 1001 | |||
| GenBank mRNA | BC159410.1 | - | 1, 2 | 707 | 8366695 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Osr1| Gene Symbol: Osr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106716.2 | NP_001100186.1 | 1, 2 | 707 | ||
| XM_006239886.2 | XP_006239948.1 | 2, 3 | 1001 | |||
| GenBank mRNA | BC159410.1 | - | 1, 2 | 707 | 8366695 | |