siRNA Details
| Gene Symbol: Nt5c1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011982.1 | NP_001011982.1 | 4 | 825 | ||
| XM_006239888.2 | XP_006239950.1 | 5 | 1027 | |||
| XM_006239889.2 | XP_006239951.1 | 5 | 1024 | |||
| XM_006239890.2 | XP_006239952.1 | 5 | 904 | |||
| XM_006239891.2 | XP_006239953.1 | 4 | 850 | |||
| XM_006239892.2 | XP_006239954.1 | 4 | 861 | |||
| GenBank mRNA | BC079437.1 | - | 4 | 825 | 7115294 | |

Nt5c1b
Genomic Map