siRNA Details
| Gene Symbol: Gen1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106717.1 | NP_001100187.1 | 13 | 1618 | ||
| XM_006239896.1 | XP_006239958.1 | 12 | 1445 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gen1| Gene Symbol: Gen1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106717.1 | NP_001100187.1 | 13 | 1618 | ||
| XM_006239896.1 | XP_006239958.1 | 12 | 1445 | |||