siRNA Details
| Gene Symbol: Cpsf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001030030.1 | NP_001025201.1 | 15 | 1833 | ||
| GenBank mRNA | BC099817.1 | - | 15 | 1833 | 7929543 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cpsf3| Gene Symbol: Cpsf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001030030.1 | NP_001025201.1 | 15 | 1833 | ||
| GenBank mRNA | BC099817.1 | - | 15 | 1833 | 7929543 | |