siRNA Details
Gene Symbol: Cpsf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001030030.1 | NP_001025201.1 | 13, 14 | 1688 | ||
GenBank mRNA | BC099817.1 | - | 13, 14 | 1688 | 7929543 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Cpsf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001030030.1 | NP_001025201.1 | 13, 14 | 1688 | ||
GenBank mRNA | BC099817.1 | - | 13, 14 | 1688 | 7929543 |