siRNA Details
| Gene Symbol: Sntg2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106720.1 | NP_001100190.1 | 10, 11 | 1159 | ||
| XM_006239967.2 | XP_006240029.1 | 14, 15 | 1440 | |||
| XM_006239968.2 | XP_006240030.1 | 13, 14 | 1320 | |||

Sntg2
Genomic Map