siRNA Details
| Gene Symbol: Dld | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199385.2 | NP_955417.1 | 12, 13 | 1467 | ||
| XM_008764590.1 | XP_008762812.1 | 12, 13 | 1484 | |||
| GenBank mRNA | BC062069.1 | - | 12, 13 | 1471 | 5599215 | |
| FM087994.1 | - | 1, 2 | 90 | |||
| FQ215299.1 | - | 12, 13 | 1473 | |||
| FQ219106.1 | - | 1, 2 | 120 | |||

Dld
Genomic Map