siRNA Details
| Gene Symbol: Scin | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198748.1 | NP_942043.1 | 12 | 1646 | ||
| XM_008764644.1 | XP_008762866.1 | 12 | 1984 | |||
| XM_008764645.1 | XP_008762867.1 | 10 | 1459 | |||
| GenBank mRNA | AY324394.1 | - | 12 | 1646 | ||

Scin
Genomic Map