siRNA Details
| Gene Symbol: Scin | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198748.1 | NP_942043.1 | 2 | 304 | ||
| XM_008764644.1 | XP_008762866.1 | 2 | 642 | |||
| GenBank mRNA | AY324394.1 | - | 2 | 304 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Scin| Gene Symbol: Scin | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198748.1 | NP_942043.1 | 2 | 304 | ||
| XM_008764644.1 | XP_008762866.1 | 2 | 642 | |||
| GenBank mRNA | AY324394.1 | - | 2 | 304 | ||