siRNA Details
Gene Symbol: Nubpl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001185025.1 | NP_001171954.1 | 4, 5 | 399 | ||
XM_008764655.1 | XP_008762877.1 | 4, 5 | 354 | |||
GenBank mRNA | BP486352.1 | - | 4, 5 | 406 | ||
CO399813.1 | - | 4, 5 | 375 | |||
FQ210674.1 | - | 4, 5 | 417 |
