siRNA Details
| Gene Symbol: Nemf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170586.1 | NP_001164057.1 | 5 | 489 | ||
| GenBank mRNA | CA512507.1 | - | 4 | 328 | ||
| CB545170.1 | - | 2 | 147 | |||
| FQ205447.1 | - | 2 | 109 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nemf| Gene Symbol: Nemf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170586.1 | NP_001164057.1 | 5 | 489 | ||
| GenBank mRNA | CA512507.1 | - | 4 | 328 | ||
| CB545170.1 | - | 2 | 147 | |||
| FQ205447.1 | - | 2 | 109 | |||