siRNA Details
| Gene Symbol: Actr10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009602.1 | NP_001009602.1 | 5 | 415 | ||
| GenBank mRNA | BC087143.1 | - | 5 | 415 | 7128051 | |
| FQ219976.1 | - | 1 | 53 | |||
| FQ228754.1 | - | 5 | 470 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Actr10| Gene Symbol: Actr10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001009602.1 | NP_001009602.1 | 5 | 415 | ||
| GenBank mRNA | BC087143.1 | - | 5 | 415 | 7128051 | |
| FQ219976.1 | - | 1 | 53 | |||
| FQ228754.1 | - | 5 | 470 | |||