siRNA Details
| Gene Symbol: RGD1565002 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271394.1 | NP_001258323.1 | 710 | |||
| GenBank mRNA | CV080419.1 | - | 5 | 670 | ||
| FM087146.1 | - | 4 | 484 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

RGD1565002| Gene Symbol: RGD1565002 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271394.1 | NP_001258323.1 | 710 | |||
| GenBank mRNA | CV080419.1 | - | 5 | 670 | ||
| FM087146.1 | - | 4 | 484 | |||