siRNA Details
| Gene Symbol: Six4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106739.1 | NP_001100209.1 | 5 | 1836 | ||
| XM_006240207.2 | XP_006240269.1 | 3 | 2000 | |||
| XM_006240208.2 | XP_006240270.1 | 4 | 2200 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Six4| Gene Symbol: Six4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106739.1 | NP_001100209.1 | 5 | 1836 | ||
| XM_006240207.2 | XP_006240269.1 | 3 | 2000 | |||
| XM_006240208.2 | XP_006240270.1 | 4 | 2200 | |||