siRNA Details
| Gene Symbol: Arel1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106744.1 | NP_001100214.1 | 12 | 1695 | ||
| XM_006240333.2 | XP_006240395.1 | 13 | 2681 | |||
| XM_006240335.2 | XP_006240397.1 | 13 | 2651 | |||
| XM_008764754.1 | XP_008762976.1 | 12 | 1956 | |||
| XM_008764755.1 | XP_008762977.1 | 14 | 2788 | |||
| GenBank mRNA | FQ226603.1 | - | 4 | 274 | ||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_178065, AK147878, BC006074, BC079590, AK147491, and others.

Arel1
Genomic Map