siRNA Details
| Gene Symbol: Dlst | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001006981.2 | NP_001006982.2 | 14 | 1231 | ||
| XM_006240306.1 | XP_006240368.1 | 12 | 1263 | |||
| XM_008764756.1 | XP_008762978.1 | 13 | 1162 | |||
| GenBank mRNA | AB504752.1 | - | 7 | 636 | ||
| BC083858.1 | - | 14 | 1217 | 7125734 | ||
| D90401.1 | - | 14 | 1182 | |||
| FQ217447.1 | - | 13 | 1156 | |||
| FQ217539.1 | - | 13 | 1156 | |||

Dlst
Genomic Map