siRNA Details
| Gene Symbol: Nek9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106747.1 | NP_001100217.1 | 2 | 459 | ||
| XM_008764757.1 | XP_008762979.1 | 2 | 483 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nek9| Gene Symbol: Nek9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106747.1 | NP_001100217.1 | 2 | 459 | ||
| XM_008764757.1 | XP_008762979.1 | 2 | 483 | |||