siRNA Details
| Gene Symbol: Serpina9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106754.1 | NP_001100224.1 | 3 | 793 | ||
| XM_008764764.1 | XP_008762986.1 | 3 | 773 | |||
| XM_008764765.1 | XP_008762987.1 | 3 | 768 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Serpina9| Gene Symbol: Serpina9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106754.1 | NP_001100224.1 | 3 | 793 | ||
| XM_008764764.1 | XP_008762986.1 | 3 | 773 | |||
| XM_008764765.1 | XP_008762987.1 | 3 | 768 | |||