siRNA Details
| Gene Symbol: Med16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106768.1 | NP_001100238.1 | 8 | 1307 | ||
| XM_006240861.2 | XP_006240923.1 | 8 | 1309 | |||
| XM_006240862.2 | XP_006240924.1 | 8 | 1306 | |||
| XM_006240864.2 | XP_006240926.1 | 8 | 1306 | |||
| XM_006240865.2 | XP_006240927.1 | 8 | 1302 | |||
| XM_006240866.2 | XP_006240928.1 | 8 | 1298 | |||

Med16
Genomic Map