siRNA Details
| Gene Symbol: LOC100909645 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006244288.1 | XP_006244350.1 | 12 | 1571 | ||
| Gene Symbol: Ankrd24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001106771.1 | NP_001100241.1 | 14 | 1988 | ||
| XM_008765093.1 | XP_008763315.1 | 14 | 1864 | |||

LOC100909645
Genomic Map