siRNA Details
Gene Symbol: Ano4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106778.1 | NP_001100248.1 | 9 | 1286 | ||
XM_008765227.1 | XP_008763449.1 | 5 | 6389 | |||
XM_008765228.1 | XP_008763450.1 | 5 | 424 | |||
XM_008765229.1 | XP_008763451.1 | 4 | 6344 | |||
XM_008765230.1 | XP_008763452.1 | 4 | 319 | |||
XM_008765231.1 | XP_008763453.1 | 9 | 2057 |
