siRNA Details
| Gene Symbol: Lta4h | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001030031.1 | NP_001025202.1 | 12, 13 | 1304 | ||
| XM_006241251.2 | XP_006241313.1 | 12, 13 | 1324 | |||
| GenBank mRNA | BC099819.1 | - | 12, 13 | 1304 | 7930840 | |

Lta4h
Genomic Map