siRNA Details
| Gene Symbol: Mgat4c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135814.1 | NP_001129286.1 | 5 | 997 | ||
| XM_008765325.1 | XP_008763547.1 | 5 | 974 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mgat4c| Gene Symbol: Mgat4c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135814.1 | NP_001129286.1 | 5 | 997 | ||
| XM_008765325.1 | XP_008763547.1 | 5 | 974 | |||