siRNA Details
| Gene Symbol: Glipr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011987.1 | NP_001011987.1 | 5, 6 | 774 | ||
| GenBank mRNA | BC089858.1 | - | 5, 6 | 774 | 7384037 | |
| FQ219892.1 | - | 5, 6 | 779 | |||
| FQ220342.1 | - | 5, 6 | 780 | |||
| FQ223414.1 | - | 5, 6 | 779 | |||
| FQ226490.1 | - | 5, 6 | 795 | |||
| FQ227021.1 | - | 5, 6 | 794 | |||
| FQ229810.1 | - | 5, 6 | 844 | |||
| FQ230706.1 | - | 5, 6 | 803 | |||

Glipr1
Genomic Map