siRNA Details
| Gene Symbol: Glipr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011987.1 | NP_001011987.1 | 6 | 836 | ||
| GenBank mRNA | BC089858.1 | - | 6 | 836 | 7384037 | |
| FQ219892.1 | - | 6 | 841 | |||
| FQ220342.1 | - | 6 | 842 | |||
| FQ223414.1 | - | 6 | 841 | |||
| FQ226490.1 | - | 6 | 857 | |||
| FQ227021.1 | - | 6 | 856 | |||
| FQ229493.1 | - | 5 | 777 | |||
| FQ229810.1 | - | 6 | 906 | |||
| FQ230706.1 | - | 6 | 865 | |||

Glipr1
Genomic Map