siRNA Details
| Gene Symbol: Eif3e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011990.1 | NP_001011990.1 | 4, 5 | 374 | ||
| XM_006250385.2 | XP_006250447.1 | 4, 5 | 416 | |||
| GenBank mRNA | BC082087.1 | - | 4, 5 | 374 | 7133833 | |
| FQ215133.1 | - | 2, 3 | 185 | |||
| FQ216598.1 | - | 3, 4 | 185 | |||
| FQ228717.1 | - | 3, 4 | 185 | |||

Eif3e
Genomic Map