siRNA Details
| Gene Symbol: Eif3h | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198751.1 | NP_942046.1 | 2 | 390 | ||
| GenBank mRNA | BC060586.1 | - | 2 | 390 | 6918037 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Eif3h| Gene Symbol: Eif3h | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198751.1 | NP_942046.1 | 2 | 390 | ||
| GenBank mRNA | BC060586.1 | - | 2 | 390 | 6918037 | |